Anti-IgA Mouse Monoclonal Antibody [clone: MARA-1] (FITC (Fluorescein Isothiocyanate))

NB600-758
102134-502EA 253.85 USD
102134-502
Anti-IgA Mouse Monoclonal Antibody [clone: MARA-1] (FITC (Fluorescein Isothiocyanate))
Mouse Monoclonal Mouse anti-Rat IgA, H Chain Antibody [FITC]. Tested Applications: Flow Cytometry. Tested Reactivity: Rat.

Type: Secondary
Antigen: IgA
Clonality: Monoclonal
Clone: MARA-1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR