Anti-Yes Mouse Monoclonal Antibody [clone: 2F3E6]

Supplier: Novus Biologicals
NBP1-47520
102177-376EA 379 USD
102177-376
Anti-Yes Mouse Monoclonal Antibody [clone: 2F3E6]
Antibodies
The Yes Antibody (2F3E6) from Novus Biologicals is a mouse monoclonal antibody to Yes. This antibody reacts with human. The Yes Antibody (2F3E6) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: Yes
Clonality: Monoclonal
Clone: 2F3E6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR