Anti-Yersinia Pestis V Antigen Mouse Monoclonal Antibody [clone: Va68]

NB100-73230
102116-226EA 244.62 USD
102116-226
Anti-Yersinia Pestis V Antigen Mouse Monoclonal Antibody [clone: Va68]
The Yersinia Pestis V Antigen Antibody (Va68) from Novus Biologicals is a mouse monoclonal antibody to Yersinia Pestis V Antigen. This antibody reacts with bacteria. The Yersinia Pestis V Antigen Antibody (Va68) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: Yersinia Pestis V Antigen
Clonality: Monoclonal
Clone: Va68
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR