Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Yersinia Pestis V Antigen Mouse Monoclonal Antibody [clone: Va13]

Supplier: Novus Biologicals
NB100-73227
The Yersinia Pestis V Antigen Antibody (Va13) from Novus Biologicals is a mouse monoclonal antibody to Yersinia Pestis V Antigen. This antibody reacts with bacteria. The Yersinia Pestis V Antigen Antibody (Va13) has been validated for the following applications: Western Blot, Sandwich ELISA.

Type: Primary
Antigen: Yersinia Pestis V Antigen
Clonality: Monoclonal
Clone: Va13
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bacteria




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
0.2 mg NB100-73227 102116-220 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol LcrV
Antigen Name Yersinia Pestis V Antigen
Reactivity Bacteria
Conjugation Unconjugated
Clonality Monoclonal
Antibody Type Primary
Clone Va13
Host Mouse
Isotype IgG1
Western Blot Yes
ELISA Yes
Antigen Synonyms V antigen LcrV, LcrV, V antigen antihost protein/regulator, Low calcium response locus protein V, Virulence associated V antigen
Immunogen Y. pestis, V antigen
Purification Protein G purified
Storage Buffer PBS (pH 7.4)
Storage Temperature Store at 4C. Do not freeze.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty