Anti-Y. pestis F1 Mouse Monoclonal Antibody

NB110-17196
102121-564EA 244.62 USD
102121-564
Anti-Y. pestis F1 Mouse Monoclonal Antibody
Mouse Monoclonal Yersinia Pestis F1 Antigen Antibody (YPF19). Tested Applications: Western Blot, ELISA, Immunohistochemistry. Tested Reactivity: Bacteria.

Type: Primary
Antigen: Y. pestis F1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR