Anti-Yellow Fever Virus Mouse Monoclonal Antibody [clone: 0G5]

Supplier: Novus Biologicals
NB100-64510
102113-862EA 564.74 USD
102113-862
Anti-Yellow Fever Virus Mouse Monoclonal Antibody [clone: 0G5]
Antibodies
The Yellow Fever Virus Antibody (0G5) from Novus Biologicals is a mouse monoclonal antibody to Yellow Fever Virus. This antibody reacts with virus. The Yellow Fever Virus Antibody (0G5) has been validated for the following applications: Immunocytochemistry / Immunofluorescence, Functional.

Type: Primary
Antigen: Yellow Fever Virus
Clonality: Monoclonal
Clone: 0G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR