Anti-XRCC3 Mouse Monoclonal Antibody (Biotin) [clone: 10F1/6]

Supplier: Novus Biologicals
NB100-180B
102106-044EA 429 USD
102106-044
Anti-XRCC3 Mouse Monoclonal Antibody (Biotin) [clone: 10F1/6]
Antibodies
The XRCC3 Antibody (10F1 / 6) [Biotin] from Novus Biologicals is a mouse monoclonal antibody to XRCC3. This antibody reacts with human, mouse (negative). The XRCC3 Antibody (10F1 / 6) [Biotin] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: XRCC3
Clonality: Monoclonal
Clone: 10F1/6
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse (Negative)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR