Anti-XPF Mouse Monoclonal Antibody [clone: 4E11]

Supplier: Novus Biologicals
NBP2-01020
102228-778EA 663.82 USD
102228-778
Anti-XPF Mouse Monoclonal Antibody [clone: 4E11]
Antibodies
The XPF Antibody (4E11) from Novus Biologicals is a mouse monoclonal antibody to XPF. This antibody reacts with human, mouse. The XPF Antibody (4E11) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: XPF
Clonality: Monoclonal
Clone: 4E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR