Anti-XPF Mouse Monoclonal Antibody [clone: XPF 3F2/3]

Supplier: Novus Biologicals
NBP1-27881
102161-250EA 299 USD
102161-250
Anti-XPF Mouse Monoclonal Antibody [clone: XPF 3F2/3]
Antibodies
The XPF Antibody (XPF 3F2 / 3) from Novus Biologicals is a mouse monoclonal antibody to XPF. This antibody reacts with human. The XPF Antibody (XPF 3F2 / 3) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: XPF
Clonality: Monoclonal
Clone: XPF 3F2/3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR