Anti-XLF Mouse Monoclonal Antibody [clone: 3D6]

Supplier: Novus Biologicals
NBP2-03275
102233-286EA 663.82 USD
102233-286
Anti-XLF Mouse Monoclonal Antibody [clone: 3D6]
Antibodies
The XLF Antibody (3D6) from Novus Biologicals is a mouse monoclonal antibody to XLF. This antibody reacts with human, monkey. The XLF Antibody (3D6) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: XLF
Clonality: Monoclonal
Clone: 3D6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR