Anti-XBP1 Mouse Monoclonal Antibody [clone: 1C4]

NBP1-47485
102177-310EA 395.38 USD
102177-310
Anti-XBP1 Mouse Monoclonal Antibody [clone: 1C4]
The XBP1 Antibody (1C4) from Novus Biologicals is a mouse monoclonal antibody to XBP1. This antibody reacts with human. The XBP1 Antibody (1C4) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: XBP1
Clonality: Monoclonal
Clone: 1C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR