Anti-WT1 Mouse Monoclonal Antibody (DyLight 488) [clone: 6F-H2]

Supplier: Novus Biologicals
NB110-60011G
102124-034EA 675.69 USD
102124-034
Anti-WT1 Mouse Monoclonal Antibody (DyLight 488) [clone: 6F-H2]
Antibodies
The WT1 Antibody (6F-H2) [DyLight 488] from Novus Biologicals is a mouse monoclonal antibody to WT1. This antibody reacts with human, mouse. The WT1 Antibody (6F-H2) [DyLight 488] has been validated for the following applications: Western Blot, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Type: Primary
Antigen: WT1
Clonality: Monoclonal
Clone: 6F-H2
Conjugation: DyLight 488
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR