Anti-VSIG2 Mouse Monoclonal Antibody [clone: 2D8]

Supplier: Novus Biologicals
NBP2-03376
102233-488EA 663.82 USD
102233-488
Anti-VSIG2 Mouse Monoclonal Antibody [clone: 2D8]
Antibodies
The VSIG2 Antibody (2D8) from Novus Biologicals is a mouse monoclonal antibody to VSIG2. This antibody reacts with human. The VSIG2 Antibody (2D8) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: VSIG2
Clonality: Monoclonal
Clone: 2D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR