Anti-Von Hippel Lindau Mouse Monoclonal Antibody [clone: 52A11]

Supplier: Novus Biologicals
NBP1-04355
102156-196EA 564.74 USD
102156-196
Anti-Von Hippel Lindau Mouse Monoclonal Antibody [clone: 52A11]
Antibodies
The Von Hippel Lindau Antibody (52A11) from Novus Biologicals is a mouse monoclonal antibody to Von Hippel Lindau. This antibody reacts with human. The Von Hippel Lindau Antibody (52A11) has been validated for the following applications: Western Blot, ELISA.

There is 1 scientific publication using this antibody.

Type: Primary
Antigen: Von Hippel Lindau
Clonality: Monoclonal
Clone: 52A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR