Anti-Vitamin D BP Mouse Monoclonal Antibody [clone: 19E8]

Supplier: Novus Biologicals
NBP1-05192
102156-682EA 379 USD
102156-682
Anti-Vitamin D BP Mouse Monoclonal Antibody [clone: 19E8]
The Vitamin D BP Antibody (19E8) - BSA Free from Novus Biologicals is a mouse monoclonal antibody to Vitamin D BP. This antibody reacts with human. The Vitamin D BP Antibody (19E8) - BSA Free has been validated for the following applications: ELISA.

Type: Primary
Antigen: Vitamin D BP
Clonality: Monoclonal
Clone: 19E8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR