Anti-Vinculin Mouse Monoclonal Antibody [clone: VIN-11-5]

Supplier: Novus Biologicals
NB120-11193
102126-802EA 564.74 USD
102126-802
Anti-Vinculin Mouse Monoclonal Antibody [clone: VIN-11-5]
Antibodies
The Vinculin Antibody (VIN-11-5) from Novus Biologicals is a mouse monoclonal antibody to Vinculin. This antibody reacts with human, mouse, bovine, chicken. The Vinculin Antibody (VIN-11-5) has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence, Microarray.

Type: Primary
Antigen: Vinculin
Clonality: Monoclonal
Clone: VIN-11-5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Bovine, Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR