Anti-Vimentin Mouse Monoclonal Antibody [clone: RV203]

Supplier: Novus Biologicals
NBP1-97671
102222-270EA 649.66 USD
102222-270
Anti-Vimentin Mouse Monoclonal Antibody [clone: RV203]
Antibodies
The Vimentin Antibody (RV203) from Novus Biologicals is a mouse monoclonal antibody to Vimentin. This antibody reacts with human, mouse, rat, porcine, canine, goat, hamster. The Vimentin Antibody (RV203) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, CyTOF-ready.

Type: Primary
Antigen: Vimentin
Clonality: Monoclonal
Clone: RV203
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Porcine, Canine, Goat, Hamster
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR