Anti-VIM Mouse Monoclonal Antibody

NBP2-11530
102249-844EA 521.54 USD
102249-844
Anti-VIM Mouse Monoclonal Antibody
Mouse Monoclonal Vimentin Antibody (33C7). Tested Applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoprecipitation. Tested Reactivity: Human, Rat.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:400

Type: Primary
Antigen: VIM
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR