Anti-Vimentin Mouse Monoclonal Antibody [clone: YT33]

Supplier: Novus Biologicals
NBP1-97524
102221-992EA 329 USD
102221-992
Anti-Vimentin Mouse Monoclonal Antibody [clone: YT33]
Antibodies
The Vimentin [p Ser33] Antibody (YT33) from Novus Biologicals is a mouse monoclonal antibody to Vimentin. This antibody reacts with human, rat. The Vimentin [p Ser33] Antibody (YT33) has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: Vimentin
Clonality: Monoclonal
Clone: YT33
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR