Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-VEGF-C Mouse Monoclonal Antibody [clone: MM0006-2E65]

Supplier: Novus Biologicals
NB110-60971
The VEGF-C Antibody (MM0006-2E65) from Novus Biologicals is a mouse monoclonal antibody to VEGF-C. This antibody reacts with human. The VEGF-C Antibody (MM0006-2E65) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:25 - 1:50

Type: Primary
Antigen: VEGF-C
Clonality: Monoclonal
Clone: MM0006-2E65
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μg NB110-60971 102124-314 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol VEGF-C
Antigen Name Vascular Endothelial Growth Factor C
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody Type Primary
Clone MM0006-2E65
Host Mouse
Isotype IgG2
Western Blot Yes
ImmunoChemistry Yes
Antigen Synonyms Flt4 ligand, vascular endothelial growth factor C, Flt4-L, VRPFLT4 ligand DHM, VEGF-C, Vascular endothelial growth factor-related protein
Gene ID 7424
Concentration LYOPH
Immunogen This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells) from a mouse immunized with recombinant human VEGF-C recombinant protein.
Purification Protein G purified
Storage Buffer A 0.2 um filtered solution in PBS
Storage Temperature Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty