Anti-VEGF-B Mouse Monoclonal Antibody [clone: MM0008-7B43]

Supplier: Novus Biologicals
NB110-60974
102124-318EA 279 USD
102124-318
Anti-VEGF-B Mouse Monoclonal Antibody [clone: MM0008-7B43]
The VEGF-B Antibody (MM0008-7B43) from Novus Biologicals is a mouse monoclonal antibody to VEGF-B. This antibody reacts with human. The VEGF-B Antibody (MM0008-7B43) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:25-1:100

Type: Primary
Antigen: VEGF-B
Clonality: Monoclonal
Clone: MM0008-7B43
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR