Anti-VEGF R3/Flt-4 Mouse Monoclonal Antibody [clone: 9D9]

Supplier: Novus Biologicals
NBP1-18651
102157-848EA 706.28 USD
102157-848
Anti-VEGF R3/Flt-4 Mouse Monoclonal Antibody [clone: 9D9]
Antibodies
The VEGF R3 / Flt-4 Antibody (9D9) from Novus Biologicals is a mouse monoclonal antibody to VEGF R3 / Flt-4. This antibody reacts with human. The VEGF R3 / Flt-4 Antibody (9D9) has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, CyTOF-ready.

Recommended Dilutions: Immunohistochemistry (Parafin): 1-10 ?g/ml

Type: Primary
Antigen: VEGF R3/Flt-4
Clonality: Monoclonal
Clone: 9D9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR