Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-VEGF R3/Flt-4 Mouse Monoclonal Antibody (Biotin) [clone: 9D9]

Supplier: Novus Biologicals
NBP1-18652
The VEGF R3 / Flt-4 Antibody (9D9) [Biotin] from Novus Biologicals is a mouse monoclonal antibody to VEGF R3 / Flt-4. This antibody reacts with human. The VEGF R3 / Flt-4 Antibody (9D9) [Biotin] has been validated for the following applications: Western Blot, ELISA, Immunoprecipitation.

Type: Primary
Antigen: VEGF R3/Flt-4
Clonality: Monoclonal
Clone: 9D9
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
0.05 mg NBP1-18652 102157-850 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol VEGF R3/Flt-4
Antigen Name FMS Related Tyrosine Kinase 4
Reactivity Human
Conjugation Biotin
Clonality Monoclonal
Antibody Type Primary
Clone 9D9
Host Mouse
Isotype IgG1
Western Blot Yes
ELISA Yes
ImmunoPrecipitation Yes
Antigen Synonyms VEGFR3Fms-like tyrosine kinase 4, VEGFR-3, soluble VEGFR3 variant 1, soluble VEGFR3 variant 2, LMPH1A, soluble VEGFR3 variant 3, EC 2.7.10.1, fms-related tyrosine kinase 4, EC 2.7.10, FLT-4, Tyrosine-protein kinase receptor FLT4, PCLFLT41, vascular endothelial growth factor receptor 3
Gene ID 2324
Concentration LYOPH
Immunogen Recombinant human soluble FLT-4 protein
Purification Protein A purified
Storage Buffer 0.1 M Tris/HCl, BSA, and 0.2 M NaCl
Storage Temperature Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty