Anti-KDR Mouse Monoclonal Antibody (Biotin)

NB100-40753
102108-842EA 521.54 USD
102108-842
Anti-KDR Mouse Monoclonal Antibody (Biotin)
Mouse Monoclonal VEGF Receptor 2 Antibody (EIC) [Biotin]. Tested Applications: Flow Cytometry. Tested Reactivity: Human, Rat.

Type: Primary
Antigen: KDR
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR