Anti-VEGF R2/KDR/Flk-1 Mouse Monoclonal Antibody [clone: 20I6]

Supplier: Novus Biologicals
NBP1-18646
102157-842EA 564.74 USD
102157-842
Anti-VEGF R2/KDR/Flk-1 Mouse Monoclonal Antibody [clone: 20I6]
Antibodies
The VEGF R2 / KDR / Flk-1 Antibody (20I6) from Novus Biologicals is a mouse monoclonal antibody to VEGF R2 / KDR / Flk-1. This antibody reacts with human. The VEGF R2 / KDR / Flk-1 Antibody (20I6) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Frozen.

Type: Primary
Antigen: VEGF R2/KDR/Flk-1
Clonality: Monoclonal
Clone: 20I6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR