Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-VE-Cadherin Mouse Monoclonal Antibody [clone: BV9]

Supplier: Novus Biologicals
The VE-Cadherin Antibody (BV9) from Novus Biologicals is a mouse monoclonal antibody to VE-Cadherin. This antibody reacts with human. The VE-Cadherin Antibody (BV9) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Functional, Immunoassay.

Type: Primary
Antigen: VE-Cadherin
Clonality: Monoclonal
Clone: BV9
Conjugation: Unconjugated
Host: Mouse
Isotype: IgG2a
Reactivity: Human

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
0.05 mg NB600-1409 102133-302 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen Symbol CDH5
Antigen Name VE-Cadherin
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody Type Primary
Clone BV9
Host Mouse
Isotype IgG2a
Western Blot Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Flow Cytometry Yes
Antigen Synonyms endothelial-specific cadherin, cadherin 5, cadherin-5, CD144 antigen, Vascular endothelial cadherin, VE-cadherin (vascular epithelium), FLJ17376, 7B4, type 2, VE-cadherin, 7B4 antigen, type 2 (vascular endothelium), CD144
Gene ID 1003
Concentration 0.1 mg/ml
Immunogen The antibody recognizes the extracellular domain of the protein.
Purification Protein A or G purified
Storage Buffer PBS with 0.1% BSA
Storage Temperature Store at 4C. Do not freeze.
Tested Applications Functional, Immunoassay
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty