Anti-VCAM1 Mouse Monoclonal Antibody

NBP1-42790
102171-618EA 395.38 USD
102171-618
Anti-VCAM1 Mouse Monoclonal Antibody
Mouse Monoclonal VCAM1 Antibody (STA). Tested Applications: ELISA, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Immunoprecipitation. Tested Reactivity: Human.

Type: Primary
Antigen: VCAM1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR