Anti-VCAM1 Mouse Monoclonal Antibody

NBP1433680.1MG NBP1433680.5MG
102172-608EA 521.54 USD
102172-608 102172-606
Anti-VCAM1 Mouse Monoclonal Antibody
Mouse Monoclonal VCAM1 Antibody (MR106). Tested Applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Immunoprecipitation. Tested Reactivity: Rat.

Type: Primary
Antigen: VCAM1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR