Anti-VCAM1 Mouse Monoclonal Antibody [clone: 6G9]

Supplier: Novus Biologicals
NBP1-47491
102177-322EA 491.08 USD
102177-322
Anti-VCAM1 Mouse Monoclonal Antibody [clone: 6G9]
Antibodies
The VCAM-1 / CD106 Antibody (6G9) from Novus Biologicals is a mouse monoclonal antibody to VCAM-1 / CD106. This antibody reacts with human. The VCAM-1 / CD106 Antibody (6G9) has been validated for the following applications: Western Blot, Simple Western, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:200 - 1:1000

Type: Primary
Antigen: VCAM1
Clonality: Monoclonal
Clone: 6G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR