Anti-VCAM1 Mouse Monoclonal Antibody [clone: 3H10]

Supplier: Novus Biologicals
NBP2-03600
102233-936EA 663.82 USD
102233-936
Anti-VCAM1 Mouse Monoclonal Antibody [clone: 3H10]
Antibodies
The VCAM-1 / CD106 Antibody (3H10) from Novus Biologicals is a mouse monoclonal antibody to VCAM-1 / CD106. This antibody reacts with human, rat, canine, monkey. The VCAM-1 / CD106 Antibody (3H10) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: VCAM1
Clonality: Monoclonal
Clone: 3H10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat, Canine, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR