Anti-VASA Mouse Monoclonal Antibody [clone: 2F9H5]

Supplier: Novus Biologicals
NBP1-51586
102181-892EA 564.74 USD
102181-892
Anti-VASA Mouse Monoclonal Antibody [clone: 2F9H5]
Antibodies
The VASA Antibody (2F9H5) from Novus Biologicals is a mouse monoclonal antibody to VASA. This antibody reacts with human. The VASA Antibody (2F9H5) has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:200 - 1:1000

Type: Primary
Antigen: VASA
Clonality: Monoclonal
Clone: 2F9H5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR