Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Varicella Zoster Virus Mouse Monoclonal Antibody [clone: NCP-1]

Supplier: Novus Biologicals
NB600-1248
The Varicella Zoster Virus Antibody (NCP-1) from Novus Biologicals is a mouse monoclonal antibody to Varicella Zoster Virus. This antibody reacts with primate, virus. The Varicella Zoster Virus Antibody (NCP-1) has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation.

Type: Primary
Antigen: Varicella Zoster Virus
Clonality: Monoclonal
Clone: NCP-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Primate, Virus




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
0.125 mg NB600-1248 102133-108 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol VZV
Antigen Name Varicella Zoster Virus
Reactivity Primate, Virus
Conjugation Unconjugated
Clonality Monoclonal
Antibody Type Primary
Clone NCP-1
Host Mouse
Isotype IgG1
ImmunoChemistry Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Antigen Synonyms VZVgI, GPIV, Glycoprotein IV, Membrane glycoprotein gE, Gi, Envelope glycoprotein gI, VZVgE, VZV, HHV3, Human herpesvirus 3, Human herpes virus 3, HHV3gp39, HHV 3
Concentration 1.05 mg/ml
Immunogen VZV Ellen Strain from VZV-infected monkey kidney cells (BSC-1).
Purification Protein G purified
Storage Buffer 20mM Sodium Phosphate (pH 9.0)
Storage Temperature Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty