Anti-VAMP3 Mouse Monoclonal Antibody

NBP19794850UL NBP197948100UL
102222-816EA 395.38 USD
102222-816 102222-818
Anti-VAMP3 Mouse Monoclonal Antibody
mouse Monoclonal VAMP3/Cellubrevin Antibody (k2A2). Tested Applications: Western Blot, ELISA. Tested Reactivity: Human, Mouse.

Type: Primary
Antigen: VAMP3
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR