Anti-VAMP1 Mouse Monoclonal Antibody [clone: SP11]

NB100-63292
102112-400EA 423.08 USD
102112-400
Anti-VAMP1 Mouse Monoclonal Antibody [clone: SP11]
The VAMP-1 Antibody (SP11) from Novus Biologicals is a mouse monoclonal antibody to VAMP-1. This antibody reacts with human, porcine, amphibian, rat (negative). The VAMP-1 Antibody (SP11) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: VAMP1
Clonality: Monoclonal
Clone: SP11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Porcine, Amphibian, Rat (Negative)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR