Anti-Vaccinia Virus Mouse Monoclonal Antibody [clone: TV46]

Supplier: Novus Biologicals
NB110-17317
102121-576EA 279 USD
102121-576
Anti-Vaccinia Virus Mouse Monoclonal Antibody [clone: TV46]
The Vaccinia Virus Antibody (TV46) from Novus Biologicals is a mouse monoclonal antibody to Vaccinia Virus. This antibody reacts with virus. The Vaccinia Virus Antibody (TV46) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: Vaccinia Virus
Clonality: Monoclonal
Clone: TV46
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR