Anti-V5 Tag Mouse Monoclonal Antibody

NBP1-21646
102160-378EA 423.08 USD
102160-378
Anti-V5 Tag Mouse Monoclonal Antibody
Mouse Monoclonal V5 Epitope Tag Antibody (SV5-Pk3). Tested Applications: Western Blot, ELISA, Immunoprecipitation. Tested Reactivity: Non-species specific.

Type: Primary
Antigen: V5 Tag
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Many Species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR