Anti-SCGB1A1 Mouse Monoclonal Antibody [clone: AY1E6]

Supplier: Novus Biologicals
NBP1-66576
102196-008EA 249 USD
102196-008
Anti-SCGB1A1 Mouse Monoclonal Antibody [clone: AY1E6]
Antibodies
The Uteroglobin / SCGB1A1 Antibody (AY1E6) from Novus Biologicals is a mouse monoclonal antibody to Uteroglobin / SCGB1A1. This antibody reacts with human. The Uteroglobin / SCGB1A1 Antibody (AY1E6) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoassay.

Type: Primary
Antigen: SCGB1A1
Clonality: Monoclonal
Clone: AY1E6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR