Anti-USP9x Mouse Monoclonal Antibody [clone: 2G7]

Supplier: Novus Biologicals
NBP2-03824
102234-384EA 663.82 USD
102234-384
Anti-USP9x Mouse Monoclonal Antibody [clone: 2G7]
Antibodies
The USP9x Antibody (2G7) from Novus Biologicals is a mouse monoclonal antibody to USP9x. This antibody reacts with human, mouse, rat, canine, primate. The USP9x Antibody (2G7) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: USP9x
Clonality: Monoclonal
Clone: 2G7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat, Canine, Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR