Anti-USP7 Mouse Monoclonal Antibody [clone: 5F11]

Supplier: Novus Biologicals
NBP1-51598
102181-916EA 329 USD
102181-916
Anti-USP7 Mouse Monoclonal Antibody [clone: 5F11]
Antibodies
The USP7 Antibody (5F11) from Novus Biologicals is a mouse monoclonal antibody to USP7. This antibody reacts with human. The USP7 Antibody (5F11) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: USP7
Clonality: Monoclonal
Clone: 5F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR