Anti-USP13 Mouse Monoclonal Antibody [clone: 4G11]

Supplier: Novus Biologicals
NBP1-48047
102178-326EA 663.82 USD
102178-326
Anti-USP13 Mouse Monoclonal Antibody [clone: 4G11]
Antibodies
The USP13 Antibody (4G11) from Novus Biologicals is a mouse monoclonal antibody to USP13. This antibody reacts with human, canine, primate. The USP13 Antibody (4G11) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: USP13
Clonality: Monoclonal
Clone: 4G11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Canine, Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR