Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-u-Plasminogen Activator/Urokinase Mouse Monoclonal Antibody [clone: PGM2005]

Supplier: Novus Biologicals
NBP1-42219
The u-Plasminogen Activator / Urokinase Antibody (PGM2005) from Novus Biologicals is a mouse monoclonal antibody to u-Plasminogen Activator / Urokinase. This antibody reacts with human, rat. The u-Plasminogen Activator / Urokinase Antibody (PGM2005) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: u-Plasminogen Activator/Urokinase
Clonality: Monoclonal
Clone: PGM2005
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
1 ml NBP1-42219 102170-824 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol URK
Antigen Name u-Plasminogen Activator/Urokinase
Reactivity Human, Rat
Conjugation Unconjugated
Clonality Monoclonal
Antibody Type Primary
Clone PGM2005
Host Mouse
Isotype IgG1
Western Blot Yes
ELISA Yes
Antigen Synonyms urinary, urokinase-type plasminogen activator, plasminogen activator, EC 3.4.21, uPA, U-plasminogen activator, ATF, urokinase, EC 3.4.21.73, u-PA
Gene ID 5328
Immunogen The antibody reacts with the Urokinase antigen.
Purification Unpurified
Storage Buffer Ascites
Storage Temperature Store at 4C. Do not freeze.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping