Anti-SETD7 Mouse Monoclonal Antibody [clone: 2D10]

Supplier: Novus Biologicals
NBP2-01066
102228-870EA 663.82 USD
102228-870
Anti-SETD7 Mouse Monoclonal Antibody [clone: 2D10]
Antibodies
The SETD7 Antibody (2D10) from Novus Biologicals is a mouse monoclonal antibody to SETD7. This antibody reacts with human, mouse, rat, canine, primate. The SETD7 Antibody (2D10) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SETD7
Clonality: Monoclonal
Clone: 2D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat, Canine, Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR