Anti-SDR9C7 Mouse Monoclonal Antibody [clone: 4B5]

Supplier: Novus Biologicals
NBP2-03030
102232-796EA 663.82 USD
102232-796
Anti-SDR9C7 Mouse Monoclonal Antibody [clone: 4B5]
Antibodies
The SDR9C7 Antibody (4B5) from Novus Biologicals is a mouse monoclonal antibody to SDR9C7. This antibody reacts with human. The SDR9C7 Antibody (4B5) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: SDR9C7
Clonality: Monoclonal
Clone: 4B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR