Anti-PDGF-D/SCDGFB Mouse Monoclonal Antibody [clone: 1C1]

Supplier: Novus Biologicals
NBP2-01332
102229-400EA 663.82 USD
102229-400
Anti-PDGF-D/SCDGFB Mouse Monoclonal Antibody [clone: 1C1]
Antibodies
The PDGF-D / SCDGFB Antibody (1C1) from Novus Biologicals is a mouse monoclonal antibody to PDGF-D / SCDGFB. This antibody reacts with human. The PDGF-D / SCDGFB Antibody (1C1) has been validated for the following applications: Western Blot, Flow Cytometry.

Type: Primary
Antigen: PDGF-D/SCDGFB
Clonality: Monoclonal
Clone: 1C1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR