Anti-RP105/CD180 Mouse Monoclonal Antibody [clone: MHR73-11]

NBP1-49025
102178-804EA 395.38 USD
102178-804
Anti-RP105/CD180 Mouse Monoclonal Antibody [clone: MHR73-11]
The RP105 / CD180 Antibody (MHR73-11) from Novus Biologicals is a mouse monoclonal antibody to RP105 / CD180. This antibody reacts with human, primate. The RP105 / CD180 Antibody (MHR73-11) has been validated for the following applications: Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.

Type: Primary
Antigen: RP105/CD180
Clonality: Monoclonal
Clone: MHR73-11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR