Anti-Ribonuclease Inhibitor Mouse Monoclonal Antibody [clone: 1B7]

Supplier: Novus Biologicals
NBP2-03254
102233-244EA 663.82 USD
102233-244
Anti-Ribonuclease Inhibitor Mouse Monoclonal Antibody [clone: 1B7]
Antibodies
The Ribonuclease Inhibitor Antibody (1B7) from Novus Biologicals is a mouse monoclonal antibody to Ribonuclease Inhibitor. This antibody reacts with human. The Ribonuclease Inhibitor Antibody (1B7) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Ribonuclease Inhibitor
Clonality: Monoclonal
Clone: 1B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR