Anti-RGS5 Mouse Monoclonal Antibody [clone: 1C1]

Supplier: Novus Biologicals
NBP2-00880
102228-500EA 663.82 USD
102228-500
Anti-RGS5 Mouse Monoclonal Antibody [clone: 1C1]
Antibodies
The RGS5 Antibody (1C1) from Novus Biologicals is a mouse monoclonal antibody to RGS5. This antibody reacts with human. The RGS5 Antibody (1C1) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: RGS5
Clonality: Monoclonal
Clone: 1C1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR