Anti-Retinol Binding Protein RBP Mouse Monoclonal Antibody [clone: 2H3]

Supplier: Novus Biologicals
NBP2-00637
102228-014EA 663.82 USD
102228-014
Anti-Retinol Binding Protein RBP Mouse Monoclonal Antibody [clone: 2H3]
Antibodies
The Retinol Binding Protein RBP Antibody (2H3) from Novus Biologicals is a mouse monoclonal antibody to Retinol Binding Protein RBP. This antibody reacts with human. The Retinol Binding Protein RBP Antibody (2H3) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Retinol Binding Protein RBP
Clonality: Monoclonal
Clone: 2H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR