Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Reticulum Cells Mouse Monoclonal Antibody [clone: ED10]

Supplier: Novus Biologicals
NB100-64648
102114-012
Anti-Reticulum Cells Mouse Monoclonal Antibody [clone: ED10]
The Reticulum Cells Antibody (ED10) from Novus Biologicals is a mouse monoclonal antibody to Reticulum Cells. This antibody reacts with rat. The Reticulum Cells Antibody (ED10) has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Frozen.

Type: Primary
Antigen: Reticulum Cells
Clonality: Monoclonal
Clone: ED10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat

Order Now


 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty