Anti-Reticulon 1A/B Mouse Monoclonal Antibody [clone: RNL-3]

Supplier: Novus Biologicals
NBP1-97735
102222-394EA 299 USD
102222-394
Anti-Reticulon 1A/B Mouse Monoclonal Antibody [clone: RNL-3]
Antibodies
The Reticulon 1A / B Antibody (RNL-3) from Novus Biologicals is a mouse monoclonal antibody to Reticulon 1A / B. This antibody reacts with human, primate, rabbit. The Reticulon 1A / B Antibody (RNL-3) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Frozen, CyTOF-ready.

Type: Primary
Antigen: Reticulon 1A/B
Clonality: Monoclonal
Clone: RNL-3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Primate, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR